Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHDC8A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569100
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569100 Supplier Invitrogen™ Supplier No. PA569100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.

This gene encodes a kelch domain-containing protein which is upregulated in cancer. Upregulated expression of the encoded protein may provide an alternative pathway for tumors to maintain aggressiveness in the absence of epidermal growth factor receptor dependence. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHDC8A
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KLHDC8A
Gene Accession No. Q8IYD2
Gene Alias A630065K24Rik; kelch domain containing 8A; kelch domain-containing protein 8A; KLHDC8A; RGD1305132; S-delta-E1; substitute for delta-EGFR expression 1
Gene Symbols KLHDC8A
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human KLHDC8A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55220
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.