Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KLHDC8A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113120
Description
KLHDC8A Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a kelch domain-containing protein which is upregulated in cancer. Upregulated expression of the encoded protein may provide an alternative pathway for tumors to maintain aggressiveness in the absence of epidermal growth factor receptor dependence. Alternative splicing results in multiple transcript variants.
Specifications
| KLHDC8A | |
| Polyclonal | |
| Unconjugated | |
| KLHDC8A | |
| A630065K24Rik; kelch domain containing 8A; kelch domain-containing protein 8A; KLHDC8A; RGD1305132; S-delta-E1; substitute for delta-EGFR expression 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 213417, 55220 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.54 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8IYD2, Q91XA8 | |
| KLHDC8A | |
| Recombinant Human Kelch domain-containing protein 8A protein (1-350AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction