Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHDC8B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552566
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552566 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552566 Supplier Invitrogen™ Supplier No. PA552566
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GTVAHQDGHL LVLGGCGRAG LPLDTAETLD MASHTWLALA PLPTARAGAA AVVLGKQVLV VGGVDEVQSP VAAVEAFLMD EGRWERRATL PQAAMGVATV ERDGMVYALG Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHDC8B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KLHDC8B
Gene Accession No. Q8IXV7
Gene Alias 4931406O17Rik; CHL; FLJ11302; FP17659; kelch domain containing 8B; kelch domain-containing protein 8B; KLHDC8B; MGC35097
Gene Symbols KLHDC8B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KLHDC8B. Recombinant protein control fragment (Product #RP-89742).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200942
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.