Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHDC9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556911
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556911 Supplier Invitrogen™ Supplier No. PA556911
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VDGRWLCVVG GWDGSRRLAT VTALDTERGV WEAWTGTPGD CPPAGLSSHT CTRISDRELQ VAGREGGIHT QRRYGSIYTL RLDPSARTY Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 80%.

KLHDC9 gene ontology annotations related to this gene include cyclin binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHDC9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KLHDC9
Gene Accession No. Q8NEP7
Gene Alias 1190002J23Rik; AA087357; ESTM31; KARCA1; kelch domain containing 9; kelch domain-containing protein 9; kelch domain-containing protein 9; LOW QUALITY PROTEIN: kelch domain-containing protein 9; kelch/ankyrin repeat containing cyclin A1 interacting protein; kelch/ankyrin repeat-containing cyclin A1-interacting protein; Klhdc9; RGD1561025
Gene Symbols KLHDC9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KLHDC9. Recombinant protein control fragment (Product #RP-94406).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126823
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.