Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL1 Monoclonal Antibody (3A8)
GREENER_CHOICE

Catalog No. PIMA515700
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515700 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515700 Supplier Invitrogen™ Supplier No. MA515700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15700 targets KLHL1 in indirect ELISA, IF and WB applications and shows reactivity with Human, Mouse and Rat samples. The MA5-15700 immunogen is purified recombinant fragment of human KLHL1 expressed in E. Coli. MA5-15700 detects KLHL1 which has a predicted molecular weight of approximately 82kDa. In Western blot, MA5-15700 did not detect KLHL1 in whole rat brain lysates, but detected KLHL1 in hippocampal neuronal cell lysates.

The mammalian Kelch-like 1 (KLHL1) was initially discovered as a homolog to the Drosophila Kelch gene that is highly expressed in several brain tissues. The predicted protein domain structure of KLHL1 is characteristic of a number of proteins that bind actin, form dimers, and often act as actin-organizing proteins. Based on the presence of anti-sense RNA that spans the transcription and translation start sites as well as the first splice site of KLHL1 in brain tissue of individuals suffering from the neurodegenerative disorder spinocerebellar ataxia type 8 (SCA8), it has been suggested that KLHL1 is involved this disease and that regulation of KLHL1 protein may be affected by antisense RNA expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3A8
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene KLHL1
Gene Accession No. Q9JI74, Q9NR64
Gene Alias FLJ30047; kelch like family member 1; kelch-l like; kelch-like 1; kelch-like family member 1; kelch-like protein 1; KIAA1490; Klhl1; Mayven-related protein 2; mKIAA1490; MRP2; RP11-394C23.1
Gene Symbols KLHL1
Host Species Mouse
Immunogen Purified recombinant fragment of human KLHL1 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 290426, 57626, 93688
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.