Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564328
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564328 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564328 Supplier Invitrogen™ Supplier No. PA564328
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AVGGFDGANR LRSAEAYSPV ANTWRTIPTM FNPRSNFGIE VVDDLLFVVG GFNGFTTTFN VECYDEKTDE WYDAHDMSIY RSALSCCVVP GLA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL10
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KLHL10
Gene Accession No. Q6JEL2
Gene Alias 4921517C11Rik; kelch like family member 10; kelch-like 10; kelch-like family member 10; kelch-like protein 10; Klhl10; SPGF11; testicular tissue protein Li 104
Gene Symbols KLHL10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KLHL10. Recombinant protein control fragment (Product #RP-103970).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317719
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.