Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109886
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109886 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109886 Supplier Invitrogen™ Supplier No. PA5109886
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASLQVLEMES METAAAGSAG LAAEVRGSGT VDFGPGPGIS AMEASGGDPG PEAEDFECSS HCSELSWRQN EQRRQGLFCD ITLCFGGAGG REFRAHRSVL AAATEYFTPL LSGQFSESRS GRVEMRKWSS EPGPEPDTVE AVIEYMYTGR IRVSTGSVHE VLELADRFLL IRLKEFCGEF LKKKLHLSNC VAIHSLAHMY TLSQLALKAA DMIRRNFHKV IQDEEFYTLP FHLIRDWLSD LEITVDSEEV LFETVLKWVQ RNAEERERYF EELFKLLRLS QMKPT.

Kelch-like protein 11, Subcellular location Secreted.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL11
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KLHL11
Gene Accession No. Q8CE33
Gene Alias BC011167; FLJ10572; kelch like family member 11; kelch-like 11; kelch-like family member 11; kelch-like protein 11; KLHL11
Gene Symbols KLHL11
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 16-300 of human KLHL11 (NP_060613.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217194, 287706
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.