Learn More
Invitrogen™ KLHL11 Polyclonal Antibody

Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: YSIGGHGNFS PGFKDVTVYN PELDKWHNLE SAPKILRDVK ALAIEDRFVY IAARTPVDRD TEDGLKAVIT CYDTETRQWQ DVESLPLIDN YCFFQMSVVN S Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 99%.
Specifications
Specifications
| Antigen | KLHL11 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | KLHL11 |
| Gene Accession No. | Q9NVR0 |
| Gene Alias | BC011167; FLJ10572; kelch like family member 11; kelch-like 11; kelch-like family member 11; kelch-like protein 11; KLHL11 |
| Gene Symbols | KLHL11 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.