Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLHL12/C3IP1 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149927APC
Description
KLHL12/C3IP1 Polyclonal antibody specifically detects KLHL12/C3IP1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)Specifications
KLHL12/C3IP1 | |
Polyclonal | |
Immunoprecipitation, Western Blot (Negative) | |
C3IP1kelch-like protein 12, CUL3-interacting protein 1, DKIR, FLJ27152, kelch-like 12 (Drosophila), kelch-like protein C3IP1 | |
The immunogen for this product maps to a region between residue 150 and 200 of human Kelch-Like 12 using the numbering given in entry NP_067646.1 (GeneID 59349). | |
0.1 mL | |
DNA Repair, DNA replication Transcription Translation and Splicing | |
59349 | |
Store at 4C in the dark. | |
IgG |
Immunoprecipitation | |
APC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction