Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLHL12/C3IP1 Antibody (OTI4E1), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272118AF405
Description
KLHL12/C3IP1 Monoclonal specifically detects KLHL12/C3IP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
KLHL12/C3IP1 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
KLHL12 | |
Recombinant protein fragment corresponding to amino acids 177-405 of human KLHL12/C3IP1 (NP_067646) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI4E1 | |
Western Blot, Immunohistochemistry | |
C3IP1kelch-like protein 12, CUL3-interacting protein 1, DKIR, FLJ27152, kelch-like 12 (Drosophila), kelch-like protein C3IP1 | |
Mouse | |
Affinity Purified | |
DNA Repair, DNA replication Transcription Translation and Splicing | |
59349.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction