Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL12 Monoclonal Antibody (2G11D1)
GREENER_CHOICE

Catalog No. PIMA549300
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549300 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549300 Supplier Invitrogen™ Supplier No. MA549300
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human PC-3 whole cell, human 293T whole cell, human Hela whole cell, human Jurkat whole cell, rat brain tissue, rat PC-12 whole cell, mouse brain tissue, mouse RAW2647 whole cell. IHC: human liver cancer tissue, human thyroid cancer tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Serves as a substrate-specific adapter for the CUL3-based ubiquitin-protein E3 ligase complex. Negatively regulates the Wnt signaling pathway via the targeted ubiquitination and subsequent proteolysis of DVL3.Tissue specificity: Highly expressed in testis and at lower levels in the submandibular salivary gland.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2G11D1
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene Klhl12
Gene Accession No. Q53G59, Q8BZM0, Q8R2H4
Gene Alias C3IP1; CUL3-interacting protein 1; DKIR; DKIR homolog; FLJ27152; hDKIR; kelch like family member 12; kelch-like 12; kelch-like family member 12; kelch-like protein 12; kelch-like protein C3IP1; Klhl12; si:dkeyp-53e12.5; Unknown (protein for MGC:134560); wu:fi60g03; zgc:92570
Gene Symbols Klhl12
Host Species Mouse
Immunogen E.coli-derived human KLHL12 recombinant protein (Position: R27-I331).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 240756, 266772, 59349
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.