Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531278
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531278 Supplier Invitrogen™ Supplier No. PA531278
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (87%), Xenopus laevis (90%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Serves as a substrate-specific adapter for the CUL3-based ubiquitin-protein E3 ligase complex. Negatively regulates the Wnt signaling pathway via the targeted ubiquitination and subsequent proteolysis of DVL3.Tissue specificity: Highly expressed in testis and at lower levels in the submandibular salivary gland.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL12
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene Klhl12
Gene Accession No. Q53G59
Gene Alias C3IP1; CUL3-interacting protein 1; DKIR; DKIR homolog; FLJ27152; hDKIR; kelch like family member 12; kelch-like 12; kelch-like family member 12; kelch-like protein 12; kelch-like protein C3IP1; Klhl12; si:dkeyp-53e12.5; Unknown (protein for MGC:134560); wu:fi60g03; zgc:92570
Gene Symbols Klhl12
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human C3IP1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59349
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.