Learn More
Invitrogen™ KLHL12 Polyclonal Antibody

Description
Immunogen sequence: FAETHNCVDLM QAAEVFSQKH FPEVVQHEEF ILLSQGEVEK LIKCDEIQVD SEEPV Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
Specifications
Specifications
| Antigen | KLHL12 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Klhl12 |
| Gene Accession No. | Q53G59 |
| Gene Alias | C3IP1; CUL3-interacting protein 1; DKIR; DKIR homolog; FLJ27152; hDKIR; kelch like family member 12; kelch-like 12; kelch-like family member 12; kelch-like protein 12; kelch-like protein C3IP1; Klhl12; si:dkeyp-53e12.5; Unknown (protein for MGC:134560); wu:fi60g03; zgc:92570 |
| Gene Symbols | Klhl12 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.