Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL13 Monoclonal Antibody (8D1)
GREENER_CHOICE

Catalog No. PIMA515658
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515658 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515658 Supplier Invitrogen™ Supplier No. MA515658
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15658 targets KLHL13 in indirect ELISA, FACS, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15658 immunogen is purified recombinant fragment of human KLHL13 expressed in E. Coli. . MA5-15658 detects KLHL13 which has a predicted molecular weight of approximately 74kDa.

This gene encodes a BTB and kelch domain containing protein and belongs to the kelch repeat domain containing superfamily of proteins. The encoded protein functions as an adaptor protein that complexes with Cullin 3 and other proteins to form the Cullin 3-based E3 ubiquitin-protein ligase complex. This complex is necessary for proper chromosome segregation and completion of cytokinesis. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL13
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 8D1
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene KLHL13
Gene Accession No. Q80TF4, Q9P2N7
Gene Alias 1200009K10Rik; AI451128; Bklhd2; BTB and kelch domain containing 2; BTB and kelch domain-containing protein 2; D130072F20Rik; FLJ10262; kelch like family member 13; kelch-like 13; kelch-like family member 13; kelch-like protein 13; Kiaa1309; KLHL13; MGC74791; mKIAA1309
Gene Symbols KLHL13
Host Species Mouse
Immunogen Purified recombinant fragment of human KLHL13 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67455, 90293
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.