Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598860
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598860 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598860 Supplier Invitrogen™ Supplier No. PA598860
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KLHL13 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a BTB and kelch domain containing protein and belongs to the kelch repeat domain containing superfamily of proteins. The encoded protein functions as an adaptor protein that complexes with Cullin 3 and other proteins to form the Cullin 3-based E3 ubiquitin-protein ligase complex. This complex is necessary for proper chromosome segregation and completion of cytokinesis. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL13
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.82 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KLHL13
Gene Accession No. Q9P2N7
Gene Alias 1200009K10Rik; AI451128; Bklhd2; BTB and kelch domain containing 2; BTB and kelch domain-containing protein 2; D130072F20Rik; FLJ10262; kelch like family member 13; kelch-like 13; kelch-like family member 13; kelch-like protein 13; Kiaa1309; KLHL13; MGC74791; mKIAA1309
Gene Symbols KLHL13
Host Species Rabbit
Immunogen Recombinant Human Kelch-like protein 13 protein (1-260AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90293
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.