Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLHL14 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $499.50
Specifications
Antigen | KLHL14 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17945720
![]() |
Novus Biologicals
NBP17945720UL |
20 μL |
Each for $158.00
|
|
|||||
NBP179457
![]() |
Novus Biologicals
NBP179457 |
100 μL |
Each for $499.50
|
|
|||||
Description
KLHL14 Polyclonal specifically detects KLHL14 in Human samples. It is validated for Western Blot.Specifications
KLHL14 | |
Polyclonal | |
Rabbit | |
NP_065856 | |
57565 | |
Synthetic peptide directed towards the N terminal of human KLHL14The immunogen for this antibody is KLHL14. Peptide sequence GVGGQDGLGAPKDQQQPPQQQPSQQQQPPPQEEPGTPSSSPDDKLLTSPR. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
kelch-like 14 (Drosophila), kelch-like protein 14, KIAA1384, printor | |
KLHL14 | |
IgG | |
71 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title