Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107155
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107155 Supplier Invitrogen™ Supplier No. PA5107155
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 87%; Dog: 93%; Horse: 82%; Human: 100%; Pig: 81%; Rabbit: 93%; Rat: 81%; Yeast: 91% Immunogen sequence: GVGGQDGLGA PKDQQQPPQQ QPSQQQQPPP QEEPGTPSSS PDDKLLTSPR.

KLHL14 is a member of the KLHL family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL14
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene KLHL14
Gene Accession No. Q9P2G3
Gene Alias 6330403N15Rik; AI785250; kelch like family member 14; kelch-like 14; kelch-like family member 14; kelch-like protein 14; Kiaa1384; KLHL14; Printor; protein interactor of Torsin-1A; protein interactor of torsinA; RGD1566178; si:dkeyp-7a3.2
Gene Symbols KLHL14
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N terminal region of human KLHL14.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57565
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.