Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595805
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595805 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595805 Supplier Invitrogen™ Supplier No. PA595805
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAGDVEGFCS SIHDTSVSAG FRALYEEGLL LDVTLVIEDH QFQAHKALLA TQSDYFRIMF TADMRERDQD KIHLKGLTAT GFSHVLQFMY YGTIELSMNT VHEILQAAMY VQLIEVVKFC CSFLLAKICL ENCAEIMRLL DDFGVNIEGV REKLDTFLLD NFVPLMSRPD FLSYLSFEKL MSYLDNDHLS RFPEIELYEA VQSWLRHDRR RWRHTDTIIQ NIRFCLMTPT SVFEKVKTSE FYRYSRQLRY EVDQALNYFQ NVHQQPLLDM KSSRIRSAKP QTTVFRGMIG HSMVNSKILL; Positive Samples: 293T, U-251MG, Mouse brain, Mouse lung, Rat lung.

KLHL15 is a member of a growing superfamily of kelch repeat-containing proteins. The kelch motif was discovered in the sequence of the Drosophila kelch ORF1 protein and forms a conserved beta-propeller tertiary structure. These kelch-like proteins have been implicated in embryogenesis and carcinogenesis through cytoskeleton organization. KLHL15 mRNA is widely expressed in many tissues and is thought to be involved in protein ubiquination and cytoskeleton organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL15
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KLHL15
Gene Accession No. A2AAX3, D3ZA50, Q96M94
Gene Alias 6330500C13Rik; epididymis secretory protein Li 305; HEL-S-305; kelch like family member 15; kelch-like 15; kelch-like family member 15; kelch-like protein 15; KIAA1677; KLHL15; RGD1563101
Gene Symbols KLHL15
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human KLHL15 (NP_0851272).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 236904, 314111, 80311
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.