Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHL18 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984019
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4019 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4019 Supplier Bioss Supplier No. BS16766R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KLHL18 is a protein coding gene.

Specifications

Antigen KLHL18
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KLHL18
Gene Accession No. O94889
Gene Alias A930041K15; AW545966; kelch like family member 18; kelch-like 18; kelch-like family member 18; Kelch-like protein 18; KIAA0795; KLHL18; OK/SW-cl.74; RGD1310570
Gene Symbols KLHL18
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KLHL18.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23276
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.