Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHL2 Antibody (OTI1G7), Biotin, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP271695B

Catalog No. NBP271695B


Only null left
Add to Cart

Description

Description

KLHL2 Monoclonal specifically detects KLHL2 in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

KLHL2
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
KLHL2
Recombinant protein fragment corresponding to amino acids 1-100 and 494-593 of human KLHL2 (NP_009177) produced in HEK293T cells.
0.1 mL
11275.0
Store at 4°C in the dark.
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
OTI1G7
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
ABP-KELCH, Actin-binding protein Mayven, kelch, kelch (Drosophila)-like 2 (Mayven), kelch-like 2, Mayven (Drosophila), kelch-like protein 2, MAV, MAYVEN
Mouse
Affinity Purified
Primary
Human, Mouse, Rat, Canine, Monkey, Primate
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.