Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHL2 Antibody (OTI1G7), PE, Novus Biologicals™
SDP

Catalog No. NBP271695PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271695PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271695PE Supplier Novus Biologicals Supplier No. NBP271695PE
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KLHL2 Monoclonal specifically detects KLHL2 in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Flow Cytometry.

Specifications

Antigen KLHL2
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1G7
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ABP-KELCH, Actin-binding protein Mayven, kelch, kelch (Drosophila)-like 2 (Mayven), kelch-like 2, Mayven (Drosophila), kelch-like protein 2, MAV, MAYVEN
Gene Symbols KLHL2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-100 and 494-593 of human KLHL2 (NP_009177) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 11275.0
Target Species Human, Mouse, Rat, Canine, Monkey, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.