Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHL2 Monoclonal Antibody (OTI6D8), TrueMAB™, OriGene
SDP

Catalog No. 502542196
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2196 30 μL
50-167-2686 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2196 Supplier OriGene Supplier No. TA501549S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KLHL2 Monoclonal antibody specifically detects KLHL2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen KLHL2
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI6D8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene KLHL2
Gene Accession No. O95198
Gene Alias 6030411N21Rik; ABP-KELCH; actin-binding protein Mayven; AU020744; kelch; kelch like family member 2; kelch-like 2, Mayven; kelch-like family member 2; kelch-like protein 2; KLHL2; MAV; MAYVEN; si:ch211-287c11.1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-100 and 494-593 of human KLHL2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11275
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.