Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLHL2 Monoclonal Antibody (OTI8B8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA501547

Description
KLHL2 (kelch-like protein 2), also known as actin-binding protein Mayven, is a 593 amino acid actin-binding protein. Highly expressed in brain, KLHL2 is thought to play a role in the organization of the actin cytoskeleton in brain cells.Specifications
KLHL2 | |
Monoclonal | |
0.74 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O95198 | |
KLHL2 | |
Human recombit protein fragment corresponding to amino acids 1-100 and 494-593 of human KLHL2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI8B8 | |
Unconjugated | |
KLHL2 | |
ABP-KELCH, MAV, MAYVEN | |
Mouse | |
Affinity Chromatography | |
RUO | |
11275 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction