Learn More
KLHL2 Mouse anti-Human, Clone: OTI10C12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501618

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
KLHL2 (kelch-like protein 2), also known as actin-binding protein Mayven, is a 593 amino acid actin-binding protein. Highly expressed in brain, KLHL2 is thought to play a role in the organization of the actin cytoskeleton in brain cells.Specifications
KLHL2 | |
Monoclonal | |
Unconjugated | |
KLHL2 | |
ABP-KELCH, MAV, MAYVEN | |
Mouse | |
Affinity Chromatography | |
RUO | |
11275 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI10C12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O95198 | |
KLHL2 | |
Human recombit protein fragment corresponding to amino acids 1-100 and 494-593 of human KLHL2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.