Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597901
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597901 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597901 Supplier Invitrogen™ Supplier No. PA597901
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KLHL2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

KLHL2 (kelch-like protein 2), also known as actin-binding protein Mayven, is a 593 amino acid actin-binding protein. Highly expressed in brain, KLHL2 is thought to play a role in the organization of the actin cytoskeleton in brain cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene KLHL2
Gene Accession No. O95198, Q8JZP3
Gene Alias 6030411N21Rik; ABP-KELCH; actin-binding protein Mayven; AU020744; kelch; kelch like family member 2; kelch-like 2, Mayven; kelch-like family member 2; kelch-like protein 2; KLHL2; MAV; MAYVEN; si:ch211-287c11.1
Gene Symbols KLHL2
Host Species Rabbit
Immunogen Recombinant Human Kelch-like protein 2 protein (1-155AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11275, 77113
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.