Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLHL20 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP238708
Description
KLHL20 Polyclonal specifically detects KLHL20 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
KLHL20 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Q9Y2M5 | |
KLHL20 | |
This antibody was developed against a recombinant protein corresponding to amino acids: NIRPGETGMDVTSRCTLGDPNKLPEGVPQPARMPYISDKHPRQTLEVINLLRKHRELCDVVLVVGAKKIYAHRVILSA | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Kelch motif containing protein, kelch-like 20 (Drosophila), kelch-like ECT2 interacting protein, Kelch-like ECT2-interacting protein, kelch-like protein 20, Kelch-like protein X, KHLHX, KLEIPRP3-383J4.3, KLHLX | |
Rabbit | |
Affinity Purified | |
RUO | |
27252 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction