Learn More
Invitrogen™ KLHL20 Polyclonal Antibody

Description
Immunogen sequence: GRDDTTELSS AERYNPRTNQ WSPVVAMTSR RSGVGLAVVN GQLMAVGGFD GTTYLKTIEV FDPDANTWRL YGGMNYRRLG GGVGVIKMTH CESHIW Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Specifications
Specifications
| Antigen | KLHL20 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | KLHL20 |
| Gene Accession No. | Q9Y2M5 |
| Gene Alias | AI504637; D930050H05Rik; kelch like family member 20; Kelch motif containing protein; kelch-like 18; kelch-like 20; Kelch-like ECT2 interacting protein; kelch-like ECT2-interacting protein; kelch-like family member 20; kelch-like protein 20; Kelch-like protein X; KHLHX; Kiaa4210; Kleip; klh20; KLHL20; KLHLX; mKIAA4210; RGD1309490; RP3-383J4.3 |
| Gene Symbols | KLHL20 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.