Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHL20 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987795
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7795 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7795 Supplier Bioss Supplier No. BS7515R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a member of the kelch family of proteins, which is characterized by a 44-56 amino acid repeat motif. The kelch motif appears in many different polypeptide contexts and contains multiple potential protein-protein contact sites. Members of this family are present both throughout the cell and extracellularly, with diverse activities.

Specifications

Antigen KLHL20
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KLHL20
Gene Accession No. Q9Y2M5
Gene Alias AI504637; D930050H05Rik; kelch like family member 20; Kelch motif containing protein; kelch-like 18; kelch-like 20; Kelch-like ECT2 interacting protein; Kelch-like ECT2-interacting protein; kelch-like family member 20; Kelch-like protein 20; Kelch-like protein X; KHLHX; Kiaa4210; KLEIP; Klhl20; KLHLX; mKIAA4210; RGD1309490; RP3-383J4.3
Gene Symbols KLHL20
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KLHL20.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27252
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.