Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL21 Monoclonal Antibody (2H4)
GREENER_CHOICE

Catalog No. PIMA515654
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515654 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515654 Supplier Invitrogen™ Supplier No. MA515654
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15654 targets KLHL21 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15654 immunogen is purified recombinant fragment of human KLHL21 expressed in E. Coli.. MA5-15654 detects KLHL21 which has a predicted molecular weight of approximately 66kDa.

Substrate-specific adapter of a BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex required for efficient chromosome alignment and cytokinesis. The BCR(KLHL21) E3 ubiquitin ligase complex regulates localization of the chromosomal passenger complex (CPC) from chromosomes to the spindle midzone in anaphase and mediates the ubiquitination of AURKB. Ubiquitination of AURKB by BCR(KLHL21) E3 ubiquitin ligase complex may not lead to its degradation by the proteasome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL21
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2H4
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene KLHL21
Gene Accession No. Q9UJP4
Gene Alias 1810045K06Rik; D330008A20; kelch like family member 21; kelch-like 21; kelch-like family member 21; kelch-like protein 21; KIAA0469; Klhl21; MGC99635; mKIAA0469; RGD1305863
Gene Symbols KLHL21
Host Species Mouse
Immunogen Purified recombinant fragment of human KLHL21 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9903
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.