Learn More
Invitrogen™ KLHL22 Monoclonal Antibody (7B10)

Description
MA5-15699 targets KLHL22 in indirect ELISA, IF and WB applications and shows reactivity with Human samples. The MA5-15699 immunogen is purified recombinant fragment of human KLHL22 expressed in E. Coli. MA5-15699 detects KLHL22 which has a predicted molecular weight of approximately 72kDa.
Specifications
Specifications
| Antigen | KLHL22 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 7B10 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.03% sodium azide |
| Gene | KLHL22 |
| Gene Accession No. | Q53GT1, Q99JN2 |
| Gene Alias | 2610318I18Rik; kelch like family member 22; KELCHL; kelch-like 22; kelch-like family member 22; kelch-like protein 22; KLHL22; RGD1306288 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.