Learn More
Invitrogen™ KLHL22 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114161
Description
Immunogen sequence: ILKNFVAFSR TDKYRQLPLE KVYSLLSSNR LEVSCETEVY EGALLYHYSL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.
KLHL22 (kelch-like protein 22) is a 634 amino acid protein that is related to the Drosophila kelch protein, which is required to maintain Actin organization in ovarian ring canals. Mutations affecting Kelch function result in the failure of Kelch to associate with the ring canals and subsequent female sterility. Human KLHL22 protein contains six kelch repeats and one BTB (POZ) domain. The BTB (Broad-Complex, Tramtrack and Bric a brac) domain, also known as the POZ (Poxvirus and Zinc finger) domain, is an N-terminal homodimerization domain that contains multiple copies of kelch repeats and/or C2H2-type zinc fingers. Proteins that contain BTB domains are thought to be involved in transcriptional regulation via control of chromatin structure and function. There are two isoforms of KLHL22 that are produced as a result of alternative splicing events.
Specifications
| KLHL22 | |
| Polyclonal | |
| Unconjugated | |
| KLHL22 | |
| 2610318I18Rik; kelch like family member 22; KELCHL; kelch-like 22; kelch-like family member 22; kelch-like protein 22; KLHL22; RGD1306288 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 84861 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q53GT1 | |
| KLHL22 | |
| Synthetic peptide directed towards the N terminal region of human KLHL22 (aa 175-224). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.