Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLHL24 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305979DL594
Description
KLHL24 Polyclonal specifically detects KLHL24 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
KLHL24 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
DRE1, Kainate Receptor Interacting Protein For GluR6, Kainate Receptor-Interacting Protein For GluR6, Kelch-Like 24, Kelch-Like 24 (Drosophila), Kelch-Like Family Member 24, Kelch-Like Protein 24, KLHL24, KRIP6, Protein DRE1 | |
Produced in rabbits immunized with E. coli-derived Human KLHL24 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
54800 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction