Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KLHL24 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563289
Description
Immunogen sequence: MVLILGRRLN REDLGVRDSP ATKRKVFEMD PKSLTGHEFF DFSSGSSHAE NILQIFNEFR DSRLFTDVII CVEG Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
KLHL24 is specifically reduces kainate receptor-mediated currents in hippocampal neurons, most probably by modulating channel properties.
Specifications
KLHL24 | |
Polyclonal | |
Unconjugated | |
KLHL24 | |
1110046J11Rik; 4930429H24Rik; 6530402O20Rik; C85082; DRE1; kainate receptor interacting protein for GluR6; kainate receptor-interacting protein for GluR6; kelch like family member 24; kelch-like 24; kelch-like family member 24; kelch-like protein 24; Klhl24; KRIP6; Protein DRE1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54800 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6TFL4 | |
KLHL24 | |
Recombinant protein corresponding to Human KLHL24. Recombinant protein control fragment (Product #RP-101205). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction