Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHL24 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988233
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8233 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8233 Supplier Bioss Supplier No. BS8050R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KLHL24 is specifically reduces kainate receptor-mediated currents in hippocampal neurons, most probably by modulating channel properties.

Specifications

Antigen KLHL24
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KLHL24
Gene Accession No. Q6TFL4
Gene Alias DRE1; kainate receptor interacting protein for GluR6; Kainate receptor-interacting protein for GluR6; kelch like family member 24; kelch-like 24; kelch-like family member 24; Kelch-like protein 24; KLHL24; KRIP6; Protein DRE1
Gene Symbols KLHL24
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KLHL24.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54800
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.