Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLHL25 Mouse anti-Human, Clone:1B7, Abnova™
Mouse monoclonal antibody raised against recombinant human KLHL25.
Supplier: Abnova Corporation MAB17699
Specifications
KLHL25 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Flow Cytometry Immunocytochemistry Immunohistochemistry Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. |
|
KLHL25 | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG2b |
Western Blot | |
1B7 | |
kelch-like 25 (Drosophila) | |
In ascites (0.03% sodium azide) | |
ENC-2, ENC2, FLJ12587, FLJ45015 | |
Recombinant protein corresponding to human KLHL25 from E. coli. | |
64410 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction