Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHL26 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988234
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8234 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8234 Supplier Bioss Supplier No. BS8052R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KLHL26 protein function is currently unknown.

Specifications

Antigen KLHL26
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KLHL26
Gene Accession No. Q53HC5
Gene Alias C630013N10Rik; kelch like family member 26; kelch-like 26; kelch-like family member 26; Kelch-like protein 26; KLHL26; Klkl26; RGD1312034
Gene Symbols KLHL26
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KLHL26.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55295
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.