Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL28 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532034
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532034 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532034 Supplier Invitrogen™ Supplier No. PA532034
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: mouse liver. Predicted reactivity: Mouse (97%), Rat (96%), Rhesus Monkey (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

BTBD5 is a transcription factor containing BTB (POZ) domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL28
Applications Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Klhl28
Gene Accession No. Q9CR40, Q9NXS3
Gene Alias 2810440N09Rik; 4122402F11Rik; 4931401E10Rik; BTB (POZ) domain containing 5; BTB/POZ domain-containing protein 5; BTBD5; kelch like family member 28; kelch-like 28; kelch-like family member 28; kelch-like protein 28; klhl28
Gene Symbols Klhl28
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human KLHL28. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54813, 66689
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.