Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLHL28 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12906R

Description
BTBD5 is a transcription factor containing BTB (POZ) domain.Specifications
KLHL28 | |
Polyclonal | |
Unconjugated | |
Klhl28 | |
2810440N09Rik; 4122402F11Rik; 4931401E10Rik; BOS_20502; BTB (POZ) domain containing 5; BTB (POZ) domain containing 5-like; BTB/POZ domain-containing protein 5; BTBD5; kelch like family member 28; kelch-like 28; kelch-like 28 (Drosophila); kelch-like family member 28; Kelch-like protein 28; KLHL28; LOC100341808 | |
Rabbit | |
Protein A | |
RUO | |
54813 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9NXS3 | |
Klhl28 | |
KLH conjugated synthetic peptide derived from human KLHL28. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction