Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KLHL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549888
Description
The antibody detects endogenous levels of total KLHL3 protein.
Probable substrate-specific adapter of an E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
Specifications
KLHL3 | |
Polyclonal | |
Unconjugated | |
KLHL3 | |
7530408C15Rik; AI430941; EG627648; kelch like family member 3; kelch-like 3; kelch-like family member 3; kelch-like protein 3; KIAA1129; Klhl3; PHA2D; RGD1565218 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100503085, 26249 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
E0CZ16, Q9UH77 | |
KLHL3 | |
Synthesized peptide derived from N-terminal of human KLHL3. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction