Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589425
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589425 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589425 Supplier Invitrogen™ Supplier No. PA589425
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEGESVKLSS QTLIQAGDDE KNQRTITVNP AHMGKAFKVM NELRSKQLLC DVMIVAEDVE IEAHRVVLAA CSPYFCAMFT GDMSESKAKK IEIKDVDGQT LSKLIDYIYT; Positive Samples: 293T, A-549, Mouse liver; Cellular Location: Cytoplasm, cytoskeleton, cytosol.

Probable substrate-specific adapter of an E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KLHL3
Gene Accession No. E0CZ16, Q9UH77
Gene Alias 7530408C15Rik; AI430941; EG627648; kelch like family member 3; kelch-like 3; kelch-like family member 3; kelch-like protein 3; KIAA1129; Klhl3; PHA2D; RGD1565218
Gene Symbols KLHL3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human KLHL3 (NP_0591112).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100503085, 26249
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.