Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHL3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988235
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8235 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8235 Supplier Bioss Supplier No. BS8053R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Probable substrate-specific adapter of an E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.

Specifications

Antigen KLHL3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KLHL3
Gene Accession No. Q9UH77
Gene Alias 7530408C15Rik; AI430941; EG627648; kelch like family member 3; kelch-like 3; kelch-like family member 3; Kelch-like protein 3; KIAA1129; Klhl3; PHA2D; RGD1565218
Gene Symbols KLHL3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KLHL3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26249
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.