Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566457
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566457 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566457 Supplier Invitrogen™ Supplier No. PA566457
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VEVESYDPYTD SWTPVSPALK YVSNFSAAGC RGRLYLVGSS ACKYNALALQ CYNPVTDAWS VIASPFLPKY L Highest antigen sequence identity to the following orthologs - mouse 90%, rat 89%.

KLHL30 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL30
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KLHL30
Gene Accession No. Q0D2K2
Gene Alias kelch like family member 30; kelch-like 30; kelch-like family member 30; kelch-like protein 30; KLHL30
Gene Symbols KLHL30
Host Species Rabbit
Immunogen Recombinant Human KLHL30. Recombinant protein control fragment (Product #RP-107130).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 377007
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.