Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHL31 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA542577
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542577 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542577 Supplier Thermo Scientific Supplier No. PA542577

Rabbit Polyclonal Antibody

Peptide sequence: TPRGWHCAVT LSDRVYVMGG SQLGPRGERV DVLTVECYSP ATGQWSYAAP Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 93%; Rat: 100%; Zebrafish: 93%

KLHL31 Polyclonal antibody specifically detects KLHL31 in Human samples. It is validated for Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL31
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KLHL31
Gene Accession No. Q9H511
Gene Alias 9830147P19Rik; bA345L23.2; BKLHD6; BTB and kelch domain-containing protein 6; D930047P17Rik; Kbtbd1; kelch like family member 31; kelch repeat and BTB (POZ) domain containing 1; kelch repeat and BTB domain-containing protein 1; kelch repeat protein; kelchlike; kelch-like 31; kelch-like family member 31; kelch-like protein 31; Kelch-like protein Klhl; klhl; KLHL31; RGD1560540; wu:fa31h10; wu:fb95e08
Gene Symbols KLHL31
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human KLHL31
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401265
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.