Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558672
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558672 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558672 Supplier Invitrogen™ Supplier No. PA558672
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TSLSEEQIWQ LAVRWLEHNC HYQYMDELLQ YIRFGLMDVD TLHTVALSHP LVQASETATA LVNEALEYHQ SIYAQPVWQT RRTKPRFQS Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.

KLHL32 (Kelch Like Family Member 32) is a Protein Coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL32
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KLHL32
Gene Accession No. Q96NJ5
Gene Alias 6430524H05Rik; BKLHD5; BTB and kelch domain containing 5; BTB and kelch domain-containing protein 5; D4Ertd389e; dJ21F7.1; Gm1356; kelch like family member 32; kelch-like 32; kelch-like family member 32; kelch-like protein 32; KIAA1900; KLHL32; mKIAA1900; RGD1310364; UG0030H05; zgc:158866
Gene Symbols KLHL32
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KLHL32. Recombinant protein control fragment (Product #RP-95026).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114792
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.