Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580757
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580757 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580757 Supplier Invitrogen™ Supplier No. PA580757
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human KLHL32.

KLHL32 (Kelch Like Family Member 32) is a Protein Coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL32
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with no preservative
Gene KLHL32
Gene Accession No. Q96NJ5
Gene Alias 6430524H05Rik; BKLHD5; BTB and kelch domain containing 5; BTB and kelch domain-containing protein 5; D4Ertd389e; dJ21F7.1; Gm1356; kelch like family member 32; kelch-like 32; kelch-like family member 32; kelch-like protein 32; KIAA1900; KLHL32; mKIAA1900; RGD1310364; UG0030H05; zgc:158866
Gene Symbols KLHL32
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of the Human KLHL32.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114792
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.