Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLHL33 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31710425UL
Description
KLHL33 Polyclonal antibody specifically detects KLHL33 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
KLHL33 | |
Polyclonal | |
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
EC 2.3.1.48, EC 2.7.1.11, Kelch-Like 33, Kelch-Like 33 (Drosophila), Kelch-Like Family Member 33, Kelch-Like Protein 33 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: VLTFAYEGVLGPASQGDVLAAAEALGAPRVKAAAQQTCERAGNAGEDVKKPSQAEELRENLRGIELLYREGVGCDLKLE | |
25 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
123103 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction