Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLHL34 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298044AF350
Description
KLHL34 Polyclonal specifically detects KLHL34 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
KLHL34 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Kelch-Like 34, Kelch-Like Family Member 34, Kelch-Like Protein 34 | |
Produced in rabbits immunized with E. coli-derived Human KLHL34 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
257240 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction