Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL35 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558955
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558955 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558955 Supplier Invitrogen™ Supplier No. PA558955
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLFVIGGARQ GGVNTDKVQC FDPKEDRWSL RSPAPFSQRC LEAVSLEDTI YVMGGLMSKI FT Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL35
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KLHL35
Gene Accession No. Q6PF15
Gene Alias 2810406K13Rik; kelch like family member 35; kelch-like 35; kelch-like family member 35; kelch-like protein 35; KLHL35; RGD1565895
Gene Symbols KLHL35
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KLHL35. Recombinant protein control fragment (Product #RP-97364).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283212
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.