Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL36 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599792
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599792 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599792 Supplier Invitrogen™ Supplier No. PA599792
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total C16orf44.

C16orf44 is an open reading frame 44, found in chromosome 16.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL36
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KLHL36
Gene Accession No. Q8N4N3
Gene Alias C16orf44; C18H16orf44; CH73-77K24.1; hypothetical protein LOC553604; kelch like family member 36; kelch-like 36; kelch-like family member 36; kelch-like protein 36; Klhl36; LOW QUALITY PROTEIN: kelch-like protein 36; Unknown (protein for MGC:128624); zgc:110268
Gene Symbols KLHL36
Host Species Rabbit
Immunogen A synthesized peptide derived from human KLHL36(Accession Q8N4N3), corresponding to amino acid residues N462-D512.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79786
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.