Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KLHL36 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540938
Description
Peptide sequence: FCCEYLEQEV SEDNYLYLQE LASIYSLKRL DAFIDGFILN HFGTLSFTPD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 83%.
C16orf44 is an open reading frame 44, found in chromosome 16.
Specifications
KLHL36 | |
Polyclonal | |
Unconjugated | |
KLHL36 | |
C16orf44; C18H16orf44; CH73-77K24.1; hypothetical protein LOC553604; kelch like family member 36; kelch-like 36; kelch-like family member 36; kelch-like protein 36; Klhl36; LOW QUALITY PROTEIN: kelch-like protein 36; Unknown (protein for MGC:128624); zgc:110268 | |
Rabbit | |
Protein A | |
RUO | |
79786 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N4N3 | |
KLHL36 | |
Synthetic peptide directed towards the N-terminal of human C16ORF44 (aa 134-183). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction